Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Hexametaphosphate

Catalog No. 3055535KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-555-35KG 5 kg
30-555-325G 25 g
30-555-31KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 30-555-35KG Supplier Sigma Aldrich Supplier No. 3055535KG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium hexametaphosphate is an inorganic polyphosphate salt commonly used as a corrosion inhibitor, emulsifying agent and as a tooth whitening agent in dentifrice formulations.Sodium hexametaphosphate has been used as a polyphosphate anion source to prepare hydrated calcium polyphosphate powder. It can be used as a deflocculant to prepare suspensions of kaolinite and ceramic grade kaolin.Intended for the cytologic demonstration of Naphthol AS-D Chloroacetate Esterase in blood, bone marrow films or tissue touch preparations.

Specifications

CAS 68915-31-1
MDL Number MFCD00134118
Quantity 5 kg
Synonym SHMP; Calgon; Phosphate glass, water soluble; Polyphosphate sodium salt; Sodium polyphosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.