Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium hexanitrocobaltate(III), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 220.0°C |
Physical Form | Powder |
Quantity | 25 g |
Linear Formula | Na3Co(NO2)6 |
Merck Index | 15,8738 |
Solubility Information | Solubility in water: 720g/L (20°C). Other solubilities: slightly soluble in alcohol |
Formula Weight | 403.94 |
Packaging | Glass bottle |
Chemical Name or Material | Sodium hexanitrocobaltate(III) |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May intensify fire; oxidizer. Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause an allergic skin reaction. May cause |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. In case of major fire and large quantities: Evacuate area. Fight fire rem |
EINECSNumber | 237-077-7 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction