Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydride, 57-63% oil dispersion

Catalog No. p-4407893
Click to view available options
Quantity:
50 g
500 g
2 x 1 kg

CAS: 7646-69-7 | HNa | 23.998 g/mol

Sodium hydride is used to enhance the condensation reactions of carbonyl compounds through the Dieckmann condensation, Stobbe condensation, Darzens condensation and Claisen condensation. It acts as a reducing agent used to prepare diborane from boron trifluoride. It is also used in fuel cell vehicles. Further, it is used to dry some organic solvents. In addition to this, it is involved in the preparation of sulfur ylides, which is utilized for the conversion of ketones into epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-69-7
Molecular Formula HNa
Molecular Weight (g/mol) 23.998
MDL Number MFCD00003471
InChI Key BZKBCQXYZZXSCO-UHFFFAOYSA-N
Synonym sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid
PubChem CID 24758
IUPAC Name sodium;hydride
SMILES [H-].[Na+]

Specifications

Color White to Gray
Physical Form Moist Powder
Quantity 50 g
Solubility Information Soluble in molten sodium. Insoluble in ammonia,benzene,carbon tetrachloride,carbon disulfide and all organic solvents.
Formula Weight 24
Odor Odorless
Sensitivity Air and moisture sensitive
Appearance Off-white to gray moist powder
Density 0.92 g/cm3
Chemical Name or Material Sodium hydride
Hazard Category H260-H350
Precautionary Statement P201-P202-P223-P231+P232-P281-P308+P313-P335+P334-P370+P378q-P501c
DOTInformation Hazard Class: 4.3, Packing Group: I
EINECSNumber 231-587-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.