Sodium hydride, 57-63% oil dispersion, Alfa Aesar™

Manufacturer: Alfa Aesar 1343118

Quantity 50g

 View more versions of this product

Catalog No. AA1343118

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7646-69-7
    Physical Form Moist Powder
    Molecular Formula HNa
    Formula Weight 24
    MDL Number MFCD00003471
    Chemical Name or Material Sodium hydride

    Specifications

    Physical Form Moist Powder
    Formula Weight 24
    Quantity 50g
    Color Off-white to gray
    DOT Information Hazard Class: 4.3, Packing Group: I
    Density 0.92g/cm3
    EINECS Number 231-587-3
    Odor Odorless
    Molecular Formula HNa
    Appearance Off-white to gray moist powder
    MDL Number MFCD00003471
    CAS 7646-69-7
    Health Hazard 1 H260: In contact with water releases flammable gases which may ignite spontaneously.
    Chemical Name or Material Sodium hydride

    H260: In contact with water releases flammable gases which may ignite spontaneously.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit