Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydride, 60% dispersion in mineral oil, in soluble bags, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
7646-69-7,8042-47-5
HNa
24
MFCD00003471
BZKBCQXYZZXSCO-UHFFFAOYSA-N
sodium hydride,sodiumhydride,sodium hydride nah,sodium hydride oil dispersion,sodium monohydride,sodium hydride nad,sodiumhydrid,natrium hydride,sodium hydrid
24758
sodium;hydride
[H-].[Na+]
7646-69-7,8042-47-5
HNa
24
MFCD00003471
BZKBCQXYZZXSCO-UHFFFAOYSA-N
sodium hydride,sodiumhydride,sodium hydride nah,sodium hydride oil dispersion,sodium monohydride,sodium hydride nad,sodiumhydrid,natrium hydride,sodium hydrid
24758
sodium;hydride
[H-].[Na+]
Adsorbent, Reduction reagent
Linear Formula NaH
Fieser 02,382; 01,1075; 04,452; 05,610; 06,541; 07,335; 08,458; 09,427; 11,486; 12,447; 14,288; 16,307
Merck Index 15, 8760
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene
Formula Weight 24
Physical Form Tacky Powder
Percent Purity 57 to 63% (NaH)
Density 1.2000g/mL
Solubility Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene
Color Gray to Yellow
Melting Point 800.0°C
Quantity 250g
Flash Point 165°C
Chemical Name or Material Sodium hydride, 60% dispersion in mineral oil
Show More Show Less

GHS H Statement:
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement:


WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger