Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydride, 60% dispersion in mineral oil, in soluble bags, in resealable cans, Thermo Scientific Chemicals

Catalog No. p-192540
Click to view available options
Quantity:
500 g
2 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent, Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-69-7, 8042-47-5
Molecular Formula HNa
Molecular Weight (g/mol) 24
MDL Number MFCD00003471
InChI Key BZKBCQXYZZXSCO-UHFFFAOYSA-N
Synonym sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid
PubChem CID 24758
IUPAC Name sodium;hydride
SMILES [H-].[Na+]

Specifications

Melting Point 800.0°C
Color Gray to Yellow
Physical Form Tacky Powder
Quantity 500 g
Linear Formula NaH
Flash Point 165°C
Fieser 02,382; 01,1075; 04,452; 05,610; 06,541; 07,335; 08,458; 09,427; 11,486; 12,447; 14,288; 16,307
Merck Index 15,876
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene
Formula Weight 24
Percent Purity 57 to 63% (NaH)
Packaging Plastic bag in resealable tin can
Density 1.2000g/mL
Chemical Name or Material Sodium hydride, 60% dispersion in mineral oil, In soluble bags, in resealable cans
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement GHS P Statement:


WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-587-3
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.