Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydrogen Carbonate, -40 to +140 mesh, ≥95%, Honeywell™ Fluka™

Catalog No. 60046781
Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Drum with Inliner
Poly Bottle

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Color Colorless
pH 8.0 to 8.6 (at 20°C)
Physical Form Liquid
Quantity 12 kg
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Beilstein 4153970
Solubility Information 90g/L at 20°C
Formula Weight 84.01g/mol
Percent Purity ≥95%
Odor Odorless
Packaging Drum with Inliner
Density 2.16g/mL at 25°C
Chemical Name or Material Sodium hydrogencarbonate
Show More Show Less
RTECSNumber 205-633-8
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.