Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sodium/hydrogen exchanger 3 Mouse anti-Rat, Opossum, Mouse, Human, Unconjugated, Clone: 3H3, MilliporeSigma™

Catalog No. MABN181325 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1813-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1813-25 Supplier MilliporeSigma Supplier No. MABN181325UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NHE3, clone 3H3, Cat. No. MABN1813, is a highly specific mouse monoclonal antibody that targets Sodium/hydrogen exchanger 3 and has been tested for use in Immunohistochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen sodium/hydrogen exchanger 3
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3H3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Sodium/hydrogen exchanger 3;Na(+)/H(+) exchanger 3;Solute carrier family 9 member 3
Gene Symbols SLC9A3;NHE3
Host Species Mouse
Immunogen MBP-conjugated recombinant fragment corresponding to 356 amino acids from the C-terminal half of Opossum sodium/hydrogen exchanger 3.
Molecular Weight of Antigen ∽75 kDa observed; 94.77 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone CH3 detects Sodium/hydrogen exchanger 3 in human, opossum, and rat kidney cells. It targets an epitope within 356 amino acids from the C-terminal half.
Target Species Rat, Opossum, Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.