Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium hydrogen fumarate, 98%

Click to view available options
Quantity:
50g
250g
5873-57-4
C4H3NaO4
138.054
MFCD00039097
VRVKOZSIJXBAJG-TYYBGVCCSA-M
sodium hydrogen fumarate,monosodium fumarate,sodium fumarate,maleic acid, sodium salt,maleinian sodu polish,fumaric acid monosodium salt,ccris 7315,2-butenedioic acid 2z-, sodium salt,sodium e-3-carboxyacrylate,maleinian sodu
6076814
sodium;(E)-4-hydroxy-4-oxobut-2-enoate
C(=CC(=O)[O-])C(=O)O.[Na+]
5873-57-4
C4H3NaO4
138.054
MFCD00039097
VRVKOZSIJXBAJG-TYYBGVCCSA-M
sodium hydrogen fumarate,monosodium fumarate,sodium fumarate,maleic acid, sodium salt,maleinian sodu polish,fumaric acid monosodium salt,ccris 7315,2-butenedioic acid 2z-, sodium salt,sodium e-3-carboxyacrylate,maleinian sodu
6076814
sodium;(E)-4-hydroxy-4-oxobut-2-enoate
C(=CC(=O)[O-])C(=O)O.[Na+]
Quantity 50g
Formula Weight 138.06
Percent Purity 98%
Chemical Name or Material Sodium hydrogen fumarate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning