Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydrosulfite, tech., 85+%

Catalog No. AA3338122
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3338122 100 g
AA33381A1 1 kg
AA3338114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3338122 Supplier Thermo Scientific Chemicals Supplier No. 03338122

CAS: 7775-14-6 | Na2O4S2 | 174.096 g/mol

Sodium hydrosulfite is used as a reducing agent in aqueous solutions, sulfonating agent , chelating agent and decolorizing agent in organic reactions. It finds application in water treatment, gas purification, cleaning, leather, polymers, photography, and many others. It is involved in chemical enhanced oil recovery to stabilize polyacrylamide polymers against radical degradation in the presence of iron. It plays an important role to determine the iron content in soil chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-14-6
Molecular Formula Na2O4S2
Molecular Weight (g/mol) 174.096
MDL Number MFCD00011640
InChI Key JVBXVOWTABLYPX-UHFFFAOYSA-L
Synonym sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
PubChem CID 24489
ChEBI CHEBI:66870
SMILES [O-]S(=O)S(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 52°C (decomposition)
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range >85%
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 174.11
Sensitivity Moisture sensitive
Chemical Name or Material Sodium Hydrosulfite
Hazard Category H302-H319-X
Precautionary Statement P235+P410-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P501c
DOTInformation DOT Class: 4.2; Packing Group: II
EINECSNumber 231-890-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.