Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.0100 N (N/100) in Methanol, Ricca Chemical
SDP

Catalog No. p-3252324 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle

Catalog No. 74710132

Supplier: Ricca Chemical Company 7471.0132

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2, 67-56-1, 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
CAS Min % 0.09, 97.0, 1.19
CAS Max % 0.1, 100.0, 1.24
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Methanol
pH 12
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.0100Normal
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.