Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.0833 N (0.0833 Molar) Standardized, Reagents

Catalog No. 01333904
Change view
Click to view available options
:
Quantity: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-904 Quantity: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-904 Supplier Reagents Holdings Llc Supplier No. CS03210020F
Only null left
Add to Cart
Add to Cart

Sodium Hydroxide, 0.0833 N (0.0833 Molar) Standardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Hydroxide, 0.0833 N (0.0833 Molar) Standardized
pH 12.92
Color Colorless
CAS 7732-18-5, 1310-73-2
Boiling Point 100°C
Molecular Formula NaOH/H2O
Synonym Lye, Caustic soda
Quantity 20 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.0017
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.