Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.100 N (N/10) in Methanol, Ricca Chemical
SDP

Catalog No. R747200020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R747200020F 20 L Cubitainer
747232 1 L Natural Poly Bottle
74721 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R747200020F Supplier Ricca Chemical Company Supplier No. 74725
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-56-1 , 1310-73-2
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145

Specifications

Color Colorless
Assay Percent Range 0.1000 ± 0.0005 N at 20°C
Molecular Formula CH4O
Solubility Information Solubility in Water: Infinite
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Quantity 20 L
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Grade Laboratory
Odor Characteristic alcohol odor
Packaging Cubitainer
Solvent or Matrix Nonaqueous Solution
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 0.1 Normal, In Methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.