Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 1.00 N Solution, APHA, Spectrum™ Chemical
SDP

Catalog No. 18606538 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L

S-370, 1310-73-2

Spectrum™ Chemical Sodium Hydroxide, 1.00 N Solution, APHA is a standard solution of Caustic soda ideally suited for laboratory titrimetric techniques. You can rely on the quality of LabChem analytical test solutions for lot-to-lot reproducibility.

Chemical Identifiers

CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 0.0384, 0.9616
Concentration 1.00 N ± 0.03 N
Quantity 20 L
Grade Chemical Solution
Packaging Plastic
Chemical Name or Material Sodium Hydroxide Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.