Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Sodium Hydroxide, 1.00 N, Ricca Chemical
    
    
    
        
            Shop All Ricca Chemical Company Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                120 mL
                            
                        
                            
                                500 mL
                            
                        
                            
                                1 L
                            
                        
                            
                                4 L
                            
                        
                            
                                10 L
                            
                        
                            
                                20 L
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Cubitainer
                            
                        
                            
                                Natural Poly Bottle
                            
                        
                    Description
- Tested in an ISO 17025 Accredited Facility
 - Test Reports provided with Certified Values and Measurement Uncertainty
 - Tight release specifications for low lot-to-lot variability
 
Specifications
Specifications
| CAS Min % | 91%, 7.25 | 
| CAS Max % | 94%, 7.54 | 
| Quantity | 1 L | 
| Grade | Laboratory | 
| Concentration or Composition (by Analyte or Components) | 1.00N | 
| Packaging | Natural Poly Bottle | 
| Chemical Name or Material | Sodium Hydroxide | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction