Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 10.0 N Aqueous Solution, APHA, Spectrum™ Chemical
SDP

Catalog No. 18606547 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 mL
4 L
20 L

Catalog No. 18-606-547

Supplier: Spectrum Chemical Manufacturing Corporation S39520LTCU

Only null left
Add to Cart
Add to Cart

S-395, 1310-73-2

Spectrum™ Chemical Sodium Hydroxide, 10.0 N Aqueous Solution, APHA - As an APHA certified solution, this item meets the ASTM D1209 standards for use in water and waste water treatment facilities to determine the concentration of dissolved and particulate materials. Spectrum Chemical manufactured APHA grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 0.3, 0.7
Concentration 10 N ± 0.5 N
Quantity 20 L
Grade APHA
Packaging Plastic
Chemical Name or Material Sodium Hydroxide Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.