Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide, 2.50 N, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
25 mL
500 mL
1 L
4 L
10 L
20 L
55 gal.
Packaging:
25 mL HDPE Bottle
Cubitainer
Natural Poly Bottle
Poly Drum
Specifications
Specifications
Color | Colorless |
Boiling Point | Approximately 100°C |
Melting Point | Approximately 0°C |
CAS Max % | 9.27%, 90.73% |
CAS Min % | 90.73%, 9.27% |
Concentration | 2.50 N |
Concentration or Composition Notes | 2.50 Normal |
Quantity | 25 mL |
Grade | Laboratory |
Packaging | 25 mL HDPE Bottle |
Show More |
Safety and Handling
ShelfLife | 24 Months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction