Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide, 20% (w/v) (200 g/L), Ricca Chemical

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
CAS Min % | 82.0, 16.34 |
CAS Max % | 85.0, 17.01 |
Quantity | 1 L |
Percent Purity | 19.96 to 20.04% |
Grade | Laboratory |
Concentration or Composition (by Analyte or Components) | 20% (w/v) Aqueous Solution (200g/L) |
Packaging | Natural Poly Bottle |
Chemical Name or Material | Sodium Hydroxide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction