Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide, 30% (w/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
CAS Min % | 75.0, 22.69 |
CAS Max % | 78.0, 23.61 |
Percent Purity | 30% |
Grade | Laboratory |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous Solution |
pH | >13 |
Quantity | 1 L |
Concentration or Composition Notes | 30% (w/v) |
Chemical Name or Material | Sodium Hydroxide, 30% (w/v) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction