Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide, 30% w/w aq. soln.

Catalog No. AA87864AP
Click to view available options
Quantity:
100 mL
500 mL

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Sodium hydroxide, 30% w/v aq. solution is used as a chemical reactant or neutralization agent in various industries like chemical, pharmaceutical, textile, pulp and paper, metal and food industries and soap and detergent manufacturing. It is also used in the manufacture of synthetic fabrics such as rayon and also for manufacturing of food products. It is utilized for pH adjustment and formation of in situ sodium methylate in bioethanol and biodiesel processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 500 mL
UN Number UN1824
Solubility Information Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether.
Formula Weight 40
Concentration or Composition (by Analyte or Components) 30% w/w aq. soln.
Chemical Name or Material Sodium hydroxide
Hazard Category H290-H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE SOLUTION
EINECSNumber 215-185-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.