Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide, 5 N Aqueous Solution, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
1 L
4 L
20 L
4 x 4 L
6 x 1 L
55 gal.
Packaging:
Poly Bottle
Poly Drum
Description
Spectrum™ Chemical Sodium Hydroxide, 5 N Aqueous Solution is a standard solution of Caustic soda ideally suited for laboratory titrimetric techniques. You can rely on the quality of LabChem analytical test solutions for lot-to-lot reproducibility.
Specifications
Specifications
| CAS Max % | 0.17, 0.83 |
| Concentration | 5.0 N ± 0.2 N |
| Quantity | 20 L |
| Grade | Chemical Solution |
| Packaging | Poly Bottle |
| Chemical Name or Material | Sodium Hydroxide Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction