Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide, 50% (w/v), Analytical Reagent Grade, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
Specifications
Specifications
| Quantity | 1 L |
| IUPAC Name | sodium hydroxide |
| Molecular Weight (g/mol) | 40.00 |
| Percent Purity | 49.92 to 50.08% |
| Grade | Analytical |
| Concentration or Composition (by Analyte or Components) | 50% (w/v) Aqueous Solution |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Sodium Hydroxide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction