Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 50% (w/w) (760 g/L), Purified Grade, Ricca Chemical
SDP

Catalog No. 72901
Click to view available options
Quantity:
15 g
500 mL
1 L
4 L
20 L
55 gal.
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum

Nitrogen Free - Suitable for Kjeldahl Nitrogen Analysis

Chemical Identifiers

CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Quantity 4 L
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Percent Purity Min. 49.5%
Grade Purified
Concentration or Composition (by Analyte or Components) 50% (w/w) Aqueous Solution (760 g/L)
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide Solution, Nitrogen Free
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.