Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Hydroxide
SCI_ED

Catalog No. s25548c Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 kg
Packaging:
Reagent Grade
Catalog No. Quantity Packaging
S25548C 1 kg Reagent Grade
S25548 100 g Reagent Grade
S25548A 500 g Reagent Grade
S25548D 3 kg Reagent Grade
4 options
This item is not returnable. View return policy

Catalog No. S25548C

This item is not returnable. View return policy

High-quality reagent grade sodium hydroxide solid, perfect for educational use in chemistry labs.

  • Reagent Grade
  • NaOH
  • Solid form, white
  • 3 kg
  • Formula weight 40 g/mol
  • Designed for educational use; no Certificate of Analysis (C of A) available
  • Secure packaging suitable for classroom and lab settings

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color White
Linear Formula NaOH
Quantity 1 kg
Formula Weight 40
Grade Reagent
Packaging Reagent Grade
Physical Form Solid
Chemical Name or Material Sodium Hydroxide
ShelfLife 2 years
DOTInformation DOT Class 8, II, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.