Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide, ACS Reagent (Pellets), Reagents
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
50 kg
Specifications
Specifications
pH | 14 |
Color | White |
Boiling Point | 1390°C |
Melting Point | 318°C |
Quantity | 50 kg |
Grade | ACS |
Packaging | Fiber Drum |
Specific Gravity | 2.13 |
Physical Form | Solid |
Chemical Name or Material | Sodium Hydroxide, ACS Reagent (Pellets) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction