Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydrogen carbonate, ACS, 99.7-100.3%

Catalog No. p-4407849
Click to view available options
Quantity:
500 g
2 kg
10 kg

For Use As a Source of CO2, in Making Sodium Salts, Baking Powder | CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and as a pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, in baking powder, as a mild abrasive and a cleaning compound. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Color White
Physical Form Granules
Quantity 500 g
Assay Percent Range 99.7-100.3%
Beilstein 4153970
Solubility Information Soluble in water and methanol. Insoluble in ethanol.
IUPAC Name sodium hydrogen carbonate
Formula Weight 84.01
Grade ACS
Odor Odorless
Refractive Index 1.5
Density 2.159 g/cm3
Chemical Name or Material Sodium hydrogen carbonate, For ACS analysis
Show More Show Less
Hazard Category H303: May be harmful if swallowed.
EINECSNumber 205-633-8
RTECSNumber VZ0950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.