Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide concentrate, For 1 L standard solution, 0.1 M NaOH (0.1 N), Honeywell Fluka™

Catalog No. p-7122110
Click to view available options
Quantity:
1 L
6 x 1 Ea.

for 1 L standard solution, 0.1 M NaOH (0.1 N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point 100°C
CAS Min % ≥5.0000%, ≥85.0000%
CAS Max % ≤15.0000%, ≤95.0000%
Linear Formula NaOH
UN Number UN1824
Density 1.07 g/cm3
Quantity 6 x 1 Ea.
Formula Weight 40.0
Packaging Ampoule
Vapor Pressure 3 mmHg (37°C)
Physical Form Concentrate
Chemical Name or Material Sodium hydroxide concentrate, For 1 L standard solution, 0.1 M NaOH (0.1 N)
Show More Show Less
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 + P310
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Sodium hydroxide solution
Recommended Storage Room Temp
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.