Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide, pearl, 97%

Catalog No. AAB2441436
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Sodium hydroxide, pearl is an important inorganic chemical used in various industries worldwide. In the paper industry, it is utilized in pulping and bleaching processes. It plays an important role in aluminum etching, chemical analysis and in paint strippers. It is an active component in a wide range of domestic products such as pipe and drain cleaners, oven cleaners and in home-cleaning products. Other uses include cleaning of fats, cleaning of equipment, ion exchange resin regeneration, pH control, degreasing agent for cleaning metal surface, de-enameling and scrubbing of off-gases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydroxide, pearl is an important inorganic chemical used in various industries worldwide. In the paper industry, it is utilized in pulping and bleaching processes. It plays an important role in aluminum etching, chemical analysis and in paint strippers. It is an active component in a wide range of domestic products such as pipe and drain cleaners, oven cleaners and in home-cleaning products. Other uses include cleaning of fats, cleaning of equipment, ion exchange resin regeneration, pH control, degreasing agent for cleaning metal surface, de-enameling and scrubbing of off-gases.

Solubility
Soluble in water, methanol, glycerol and ethanol. Insoluble in acetone and ether.

Notes
Air sensitive and hygroscopic. It is deliquescent and readily absorbs moisture and carbon dioxide in air. Incompatible with strong oxidizing agents, strong acids and organic materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point 1390°C
Melting Point 318°C
pH 14
Quantity 500 g
UN Number UN1823
Merck Index 14,8627
Solubility Information Soluble in water,methanol,glycerol and ethanol. Insoluble in acetone and ether.
Formula Weight 40
Percent Purity 97%
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Density 2.13
Chemical Name or Material Sodium hydroxide, pearl
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE, SOLID
EINECSNumber 215-185-5
RTECSNumber WB4900000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.