Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium hydroxide solution, Volumetric, Reag. Ph. Eur., 0.1 M NaOH (0.1N), Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
5 L
10 L
6 x 1 L
Packaging:
Plastic Bottle
VOLPAC™ CONTAINER
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Boiling Point | 100°C |
| CAS Min % | >99.5000% |
| CAS Max % | >99.50%, <0.5000% |
| Linear Formula | NaOH |
| Concentration | 0.1 M NaOH (0.1N) |
| UN Number | UN1824 |
| Density | 1.000 g/cm3 |
| Quantity | 6 x 1 L |
| Formula Weight | 40 |
| Grade | Reagent Ph. Eur. |
| Show More |
Safety and Handling
| ShelfLife | 450 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Sodium hydroxide solution |
| EINECSNumber | 231-791-2 |
| Recommended Storage | Room Temp |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction