Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide Solution, 1 M, For HPCE, MilliporeSigma™ Supelco™

Catalog No. 111016646 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
100 mL
500 mL

Chemical Identifiers

MDL Number MFCD00003548

Specifications

CAS 1310-73-2
Linear Formula NaOH
Quantity 500 mL
Formula Weight 40.00
Grade HPCE
Concentration or Composition (by Analyte or Components) 0.9-1.1 M
Vapor Pressure 3 mmHg (37°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.