Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide Solution (1 N/Certified), Fisher Chemical PROMO

Catalog No. SS2661FB25 Shop All Fisherbrand Products
Click to view available options
:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric standard solution, titrated at 25°C to pH 8.6 (phenolphthalein endpoint) against NIST™ potassium acid phthalate.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997 g/mol
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, Caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide, sodium hydroxide Solution, soda, Caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name Sodium hydroxide
SMILES [OH-].[Na+]

Specifications

pH 13.3
Color Colorless
Boiling Point 100°C
Melting Point 0°C
Concentration 0.995 to 1.005 N
Quantity 1 L
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST LOT 84 L Potassium acid phthalate
Physical Form Liquid
Chemical Name or Material Sodium hydroxide Solution, 1 N
Show More Show Less
Hazard Category Signal Word: DANGER!
Hazard Statement DANGER!
Precautionary Statement

Emergency Overview: Causes burns by all exposure routes.

NFPA

Health:3

Flammability:0

Instability:0

DOTInformation DOT Class 8, : Corrosive
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.