Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide solution, ≥32%, for determination of nitrogen, ≤0.0001% N, Honeywell™

Catalog No. 60045829 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-829 4 x 5 L Poly Bottle
60-014-72 1 L Poly Bottle
60-014-73 2.5 L Poly Bottle
60-014-74 5 L Poly Bottle
60-045-830 6 x 1 L Poly Bottle
60-045-828 4 x 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-045-829 Supplier Honeywell Chemicals Supplier No. 305314X5L
Only null left
Add to Cart
Add to Cart

≥32%, for determination of nitrogen, ≤0.0001% N

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point 120°C
CAS Min % ≥50.0000%, ≥25.0000%
CAS Max % <75.0000%, <50.0000%
Linear Formula NaOH
UN Number UN1824
Density 1.350 g/cm3
Quantity 4 x 5 L
Formula Weight 40.0
Percent Purity >32%
Packaging Poly Bottle
Chemical Name or Material Sodium hydroxide solution, ≥32%, for determination of nitrogen, ≤0.0001% N
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Sodium hydroxide solution
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.