Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hydroxide Solution AR, Macron Fine Chemicals™
Shop All Avantor Products
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Poly
Specifications
Specifications
Assay Percent Range | 50% |
Quantity | 1 L |
Packaging | Poly |
Chemical Name or Material | Sodium Hydroxide Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction