Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hypochlorite, 12.5% Avl Cl-, Reagents

Catalog No. 01337006
Change view
Click to view available options
Quantity:
15 gal.
55 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-006 55 gal.
01-337-007 15 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-006 Supplier Reagents Holdings Llc Supplier No. C231905055E
Add to Cart
Add to Cart

Sodium Hypochlorite, 12.5% Avl Cl-, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7681-52-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Bleach
IUPAC Name water
SMILES O

Specifications

Color Colorless to Greenish-Yellow
pH 13
Physical Form Liquid
Quantity 55 gal.
Grade Technical
Packaging Poly Drum
Specific Gravity 1.243
Chemical Name or Material Sodium Hypochlorite, 12.5% Avl Cl-
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.