Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hypochlorite, 10-15% active chlorine

Catalog No. p-215352
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Plastic bottle
Plastic drum

Catalog No. AC219255000

Supplier: Thermo Scientific Chemicals 219255000

Only null left
Add to Cart

CAS: 7681-52-9 | ClNaO | 74.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9, 7732-18-5
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.44
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Melting Point -16.0°C
Color Yellow
Physical Form Solution
Quantity 500 mL
Linear Formula NaOCl
Fieser 01,1084; 04,456; 05,617; 06,543; 07,337; 08,461; 09,430; 10,365; 11,487; 15,293; 16,308
Merck Index 15,8762
Solubility Information Solubility in water: soluble
Formula Weight 74.44
Percent Purity ≥12%
Packaging Plastic bottle
Specific Gravity 1.2
Density 1.2000g/mL
Chemical Name or Material Sodium hypochlorite, 13% Active chlorine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Toxic to aquatic life with long lasting effects.
Contact with acids liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 231-668-3
RTECSNumber NH3486300
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.