Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hypochlorite, 11-15% available chlorine

Catalog No. AA33369A3
Click to view available options
Quantity:
500 g
2 kg
10 kg

CAS: 7681-52-9 | ClNaO | 74.439 g/mol

Sodium hypochlorite is used as a disinfectant and a bleaching agent. It is used as a destainer involved in removing of mold stains, dental stains arises by fluorosis. It finds application in laundry detergents. Its aqueous solution is used as household bleach for sanitizing smooth surfaces. In a water treatment plant, it is utilized for treating dilute cyanide waste water like electroplating wastes. It is an oxidizing agent used with phase transfer catalyst to convert alcohols into corresponding carbonyl compounds. It is an important reactant in Hofmann rearrangement to prepare primary amine from primary amide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146

Specifications

Color Yellow
Physical Form Liquid
Quantity 2 kg
Solubility Information Miscible with water.
SMILES [O-]Cl.[Na+]
IUPAC Name sodium;hypochlorite
Molecular Weight (g/mol) 74.439
Formula Weight 74.44
Density 1.2
Chemical Name or Material Sodium Hypochlorite aqueous Solution, 11-15% available chlorine
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation DOT Class: 8; Packing Group: III
EINECSNumber 231-668-3
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.