Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hypochlorite Solution, 2.5% (w/w) NaOCl, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
55 gal.
Packaging:
Natural Poly Bottle
Poly Drum
Specifications
Specifications
Color | Colorless to Green-Yellow |
CAS Min % | 97.0, 2.15 |
CAS Max % | 99.0, 2.23 |
Physical Form | Liquid |
Quantity | 4 L |
Percent Purity | 2.5% W/W |
Grade | Laboratory |
Concentration or Composition (by Analyte or Components) | 2.5% (w/w) NaOCl |
Packaging | Natural Poly Bottle |
Chemical Name or Material | Sodium Hypochlorite Solution |
Safety and Handling
ShelfLife | 6 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction