Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Hypochlorite Solution, 5% available Chlorine, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
55 gal.
Packaging:
Amber Poly Bottle
Black Poly Bottle
Poly Drum
Specifications
Specifications
Color | Colorless to Green-Yellow |
CAS Min % | 92.0, 5.88 |
CAS Max % | 96.0, 6.12 |
Physical Form | Liquid |
Quantity | 4 L |
Percent Purity | 5% W/W |
Grade | Laboratory |
Concentration or Composition (by Analyte or Components) | 5% available Chlorine |
Packaging | Black Poly Bottle |
Chemical Name or Material | Sodium Hypochlorite Solution |
Safety and Handling
ShelfLife | 6 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction