Sodium Hypochlorite Solution, 5% available Chlorine, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 7495.555

Packaging Drum
Quantity 55 gal.

 View more versions of this product

Catalog No. 7495-555

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,7681-52-9,7732-18-5
    CAS Min % 92.0,5.88
    CAS Max % 96.0,6.12
    Physical Form Liquid
    Chemical Name or Material Sodium Hypochlorite Solution

    Specifications

    Physical Form Liquid
    Color Clear, colorless to greenish-yellow
    Quantity 55 gal.
    Shelf Life 6 months
    Packaging Drum
    CAS 7732-18-5,7681-52-9,7732-18-5
    Concentration or Composition (by Analyte or Components) 5% available Chlorine
    CAS Min % 92.0,5.88
    CAS Max % 96.0,6.12
    Chemical Name or Material Sodium Hypochlorite Solution

    6 months

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit