Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Iodide
SCI_ED

Catalog No. S25554 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade

White sodium iodide powder ideal for classroom experiments in chemistry and physics.

  • Material: Sodium Iodide (NaI)
  • Color: White
  • Quantity: 500 grams
  • Assay Percent Range: Greater than 99% purity
  • Packaging: Lab Grade Powder
  • Density: 3.667 g/cm³
  • Usage: For educational purposes only, no Certificate of Analysis (C of A) available
  • Applications: Ideal for use in chemistry experiments, radiation detection studies, and physics demonstrations

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Color White
Physical Form Powder, Crystals and/or Chunks
Quantity 100 g
Assay Percent Range >99%
Linear Formula NaI
Formula Weight 149.89
Packaging Lab Grade
Density 3.6670g/cmsuperscript_3;
Chemical Name or Material Sodium Iodide
ShelfLife 1 year
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.