Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium iodide, 99.999%, (trace metal basis), extra pure

Catalog No. AC212680100
Click to view available options
Quantity:
1 g
10 g
100 g
Packaging:
Glass bottle

CAS: 7681-82-5 | INa | 149.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.89
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Boiling Point 1300.0°C
Melting Point 661.0°C
Color White
Physical Form Granular Powder
Quantity 10 g
Assay Percent Range 99.999%
Linear Formula NaI
Fieser 01,1087; 02,384; 03,267; 04,456; 06,543; 07,338; 10,365; 11,488; 12,29; 13,43; 14,89; 16,86
Merck Index 15, 8765
Solubility Information Solubility in water: 184 g/100mL (25°C). Other solubilities: soluble in alcohol, glycerol and acetone
Formula Weight 149.89
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 3.66
Density 3.6600g/mL
Chemical Name or Material Sodium iodide, Extra Pure
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid releas
EINECSNumber 231-679-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.