Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium iodide, 99+% (dry wt., water <1.0%)

Click to view available options
Quantity:
100g
500g
2500g
7681-82-5
INa
149.894
MFCD00003532
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide,ioduril,sodium monoiodide,soiodin,sodiumiodide,natrii iodidum,jodid sodny,iodure de sodium,jodid sodny czech
5238
CHEBI:33167
sodium;iodide
[Na+].[I-]
7681-82-5
INa
149.894
MFCD00003532
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide,ioduril,sodium monoiodide,soiodin,sodiumiodide,natrii iodidum,jodid sodny,iodure de sodium,jodid sodny czech
5238
CHEBI:33167
sodium;iodide
[Na+].[I-]
Applications
Sodium iodide is widely used for halide exchange (Finkelstein reaction), for example in the conversion of an alkyl chloride, allyl chloride and arylmethyl chloride into their respective iodides, which are precursors for pharmaceutical and fine chemical products. They are used to ehnance the efficiency of the formation of Wittig adducts from less reactive chlorides and bromides. Appropriate prepartions find use as a nutrient supplement. Sodium iodide is used as the precursor to the control agent in ab initio emulsion polymerization. Sodium iodide finds use in the determination of dissolved oxygen in the modified Winkler method, synthesis of the fluorescent dye coppersensor-1 (CS1) for imaging labile copper pools in biological samples, and the cleavage of esters, lactones, carbamates and ethers in combination with chlorotrimethylsilane.

Solubility
Very soluble in water and liquid ammonia. Freely soluble in acetone, formic acid, formamide, methanol, and acetonitrile. Soluble in N,N-dimethylformamide.

Notes
Deliquescent. Store in a tightly closed container. Incompatible with strong oxidizing agents and acids.
Merck Index 14,8631
Solubility Information Very soluble in water and liquid ammonia. Freely soluble in acetone,formic acid,formamide,methanol,and acetonitrile. Soluble in N,N-dimethylformamide.
Formula Weight 149.89
Percent Purity ≥99%
Density 3.67
Odor Odorless
Refractive Index 1.7745
Sensitivity Hygroscopic; Light sensitive
Boiling Point 1300°C to 1304°C
Melting Point 651°C
Quantity 100g
Chemical Name or Material Sodium iodide
Assay (dry wt.), water <1.0%
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 231-679-3

RTECSNumber : WB6475000

TSCA : Yes

Recommended Storage : Ambient temperatures