Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium iodide, ACS, 99.5% min

Catalog No. AA4019636
Click to view available options
Quantity:
100 g
500 g

Used to prevent iodine deficiency. Used as analytical reagents, in manufacture of disinfectants, antiseptics, deodorants and medicines, in baking ingredient conditioner. Also used for conversion of alkyl chlorides into alkyl iodides | CAS: 7681-82-5 | INa | 149.894 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
  • Commonly used to prevent iodine deficiency Used as analytical reagents, in manufacture of disinfectants, antiseptics, deodorants and medicines
  • Used in baking ingredient conditioner and as animal feed
  • Used for conversion of an alkyl chlorides into an alkyl iodides, such as in the treatment of thyroid cancer and hyperthyroidism and in scintillation detectors, traditionally in nuclear medicine, geophysics, nuclear physics, and environmental measurements
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Boiling Point 1300°C to 1304°C
Melting Point 651°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99.5% min.
Merck Index 14,8631
Solubility Information Soluble in water, alcohol, glycerol and acetone. Insoluble in ether.
Formula Weight 149.89
Grade ACS
Odor Odorless
Refractive Index 1.7745
Sensitivity Hygroscopic
Appearance White Powder
Density 3.67 g/cm3 at 20°C
Chemical Name or Material Sodium iodide
Show More Show Less
Hazard Category H315-H319
Hazard Statement P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a: Avoid breathing dust/fume/gas/mist/vapours/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Take off contaminated clothing and wash before reuse. Call a POISON CENTER or doctor/physician if you feel unwell. Specific treatment (see label). Store locked up. Store in a well-ventilated place. Keep container tightly closed. Dispose of contents/container in accordance with local/regional/national/international regulations.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-679-3
RTECSNumber WB6475000
TSCA Yes
storageNote1 Hygroscopic and incompatible with strong acids,oxidizing agents. light, air and moisture sensitive.
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.