Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium iodide, ACS reagent, anhydrous

Catalog No. AC419570025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC419570025 2.5 kg
AC419571000 100 g
AC419575000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419570025 Supplier Thermo Scientific Chemicals Supplier No. 419570025
Add to Cart

CAS: 7681-82-5 | INa | 149.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.89
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Boiling Point 1300.0°C
Melting Point 661.0°C
pH 6.0 to 9.0 (5% aq. soln.)
Quantity 2.5 kg
Linear Formula NaI
Fieser 01,1087; 02,384; 03,267; 04,456; 06,543; 07,338; 10,365; 11,488; 12,29; 13,43; 14,89; 16,86
Merck Index 15,8765
Solubility Information Solubility in water: 184 g/100mL (25°C). Other solubilities: soluble in alcohol, glycerol and acetone
Formula Weight 149.89
Grade ACS Reagent
Specific Gravity 3.66
Density 3.6600g/mL
Chemical Name or Material Sodium iodide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid releas
EINECSNumber 231-679-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.