Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium iodide hydrate, Puratronic™, 99.999% (metals basis)

Catalog No. p-7048558
Click to view available options
Quantity:
5 g
25 g
100 g

Sodium Iodide is used in the synthesis of dye-sensitized solar cells. Also used in the preparation of propargylamines as inhibitors of lysine specific. Demethylase. Used in photography and as analytical reagents. They are used in the measurement of the energy of gamma rays, by measuring the amplitude of pulses of light generated by electrons which are excited by the gamma rays and in the Finkelstein reaction via an SN2 pathway, detection of radiation, and in the conversion of an alkyl chloride into an alkyl iodide. Sodium Iodide is used in PCR and to produce chemokines, particularly CCL2, CXCL8, and CXC14 by stimulation of thyroid follicular cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00003532

Specifications

Physical Form Crystalline Aggregates
Quantity 5 g
Solubility Information Soluble in water (1840mg/ml at 25°C),alcohol,glycerol,and acetone. Insoluble in ether.
Formula Weight 149.89 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic™
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Sodium iodide hydrate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 231-679-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.