Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium-Iodide Symporter Mouse anti-Human, Unconjugated, Clone: ARN1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC1191100UG
Description
Anti-Sodium-Iodide Symporter, clone ARN1, Cat. No. MABC1191, is a mouse monoclonal antibody that detects Sodium/iodide cotransporter and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Specifications
Sodium-Iodide Symporter | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal antibody IgG2a in buffer containing 140 mM Tris and 180 mM Glycine (pH 7.4) without azide. | |
SLC5A5;NIS | |
KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal region of human Sodium/iodide cotransporter. | |
Protein G purified | |
RUO | |
Primary | |
Clone ARN1 is a mouse monoclonal antibody that detects Sodium/iodide cotransporter in human cells. It targets an epitope within 13 amino acids from the C-terminal region. | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
Purified |
Immunohistochemistry (Paraffin), Western Blot | |
ARN1 | |
Unconjugated | |
Sodium/iodide cotransporter;Na(+)/I(-) cotransporter;Na(+)/I(-) symporter;Solute carrier family 5 member 5 | |
Mouse | |
∽68 kDa observed; 68.67 kda calculated. Uncharacterized bands may be observed in some lysate(s). | |
100 μg | |
Apoptosis & Cancer | |
NP_000444.1 | |
Human | |
Antibodies | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction