Learn More
Sodium Iodide Symporter Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Sodium Iodide Symporter |
---|---|
Concentration | 0.77 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4300
![]() ![]() |
Proteintech Group Inc
243241AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody recognizes the mature approximately 75-100 kDa protein and a partially glycosylated 50-55 kDa protein.
Human Sodium Iodide Symporter (hNIS) is responsible for iodide concentrating ability within thyroid follicular cells. It is a membrane bound glycoprotein with 13 membrane spanning domains and 14 extra membranous domains. It may represent an autoantigen in thyroid.Specifications
Sodium Iodide Symporter | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q63008, Q92911, Q99PN0 | |
114479, 114613, 6528 | |
Sodium iodide symporter Fusion Protein Ag19504 | |
Primary | |
-20°C | |
SLC5A5 |
0.77 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
NIS, SLC5A5, Sodium iodide symporter, Sodium/iodide cotransporter | |
SLC5A5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.