Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Iodide, USP, 99-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18606600 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-600 45 kg Fiber Drum
18-606-601 5 g Amber Glass Bottle
18-606-602 25 g Amber Glass Bottle
18-606-596 125 g Amber Glass Bottle
18-606-597 500 g Amber Glass Bottle
18-606-598 2.5 kg Amber Glass Bottle
18-606-599 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-606-600 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO17545KGBL
Add to Cart
Add to Cart
Request a Quote

SO175, 7681-82-5, NaI

Spectrum™ Chemical Sodium Iodide, USP is a crystalline salt that is white and is used as a food supplement to counter and prevent iodine deficiencies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.89
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
IUPAC Name sodium iodide
SMILES [Na+].[I-]

Specifications

CAS Max % 1
Quantity 45 kg
Percent Purity 99 to 101.5%
Grade USP
Packaging Fiber Drum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.