Learn More
Sodium isopropoxide
Description
Sodium isopropoxide is used in the preparation of ibuprofen intermediate. It is involved in reactions such as polymerization of butadiene. Further, it acts as a catalyst in the manufacturing of acetone and its derivatives. In addition, it is used as a solvent and serves as an antiseptic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 70°C to 75°C |
Quantity | 25 g |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in tetrahydrofuran. |
Formula Weight | 82.08 |
Physical Form | Crystalline |
Chemical Name or Material | Sodium isopropoxide |
Safety and Handling
Hazard Category | H228-H314-H335-X |
Precautionary Statement | P210-P235+P410-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 4.2; Packaging Group: II |
EINECSNumber | 211-673-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.