Learn More
Sodium KAP Analysis Standard 1 Molar NaCl with ISA, Reagents
Specifications
Specifications
| Chemical Name or Material | Sodium KAP Analysis Standard 1 Molar NaCl with ISA |
| Packaging | Poly Bottle |
| CAS | 7732-18-5, 7647-14-5, 12125-02-9 |
| Specific Gravity | 1.0383 |
| Physical Form | Liquid |
| Technique | Qualitative Analysis of Cations and Anions |
| Color | Colorless |
| pH | 10 |
| Boiling Point | 102°C |
| Solubility | Completely miscible |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.